ENA321765784 Enamine

1-cyclopropyl-2-(piperazin-1-yl)ethan-1-one dihydrochloride

  • MDL number MFCD16653271
  • Cas Number : 1306604-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216...218
Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77909

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77909
Availability details for
Quantity
  • Availability
  • Add to Cart