ENA321765787 Enamine

5-(3-fluoro-2-methylphenyl)furan-2-carbaldehyde

  • MDL number MFCD16066210
  • Cas Number : 1251392-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78142
Availability details for
Quantity
  • Availability
  • Add to Cart