ENA321765788 Enamine

1-(dimethylamino)-4,4-dimethylpentan-3-one hydrochloride

  • MDL number MFCD00034852
  • Cas Number : 53400-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78143
Availability details for
Quantity
  • Availability
  • Add to Cart