ENA321765789 Enamine

N-methylpyridazin-3-amine

  • MDL number MFCD16767565
  • Cas Number : 45588-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78144
Availability details for
Quantity
  • Availability
  • Add to Cart