ENA321765790 Enamine

2-[(4-bromophenoxy)methyl]-1-(difluoromethyl)-1H-imidazole

  • MDL number MFCD11102617
  • Cas Number : 1152545-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-77914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77914
Availability details for
Quantity
  • Availability
  • Add to Cart