ENA321765794 Enamine

5-bromo-2-(2,2,2-trifluoroethoxy)pyridin-3-amine

  • MDL number MFCD14600577
  • Cas Number : 1249747-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77916
Availability details for
Quantity
  • Availability
  • Add to Cart