ENA321765797 Enamine

5-(oxolan-2-yl)-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD07376214
  • Cas Number : 1247732-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125...127
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78186

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78186
Availability details for
Quantity
  • Availability
  • Add to Cart