ENA321765799 Enamine

methyl 2-bromo-5-(chlorosulfonyl)-4-fluorobenzoate

  • MDL number MFCD12169483
  • Cas Number : 1155910-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78189
Availability details for
Quantity
  • Availability
  • Add to Cart