ENA321765804 Enamine

methyl 2-chloroquinoline-4-carboxylate

  • MDL number MFCD03413689
  • Cas Number : 62482-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87...89
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78196

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78196
Availability details for
Quantity
  • Availability
  • Add to Cart