ENA321765806 Enamine

2-(2-{[(2,2,2-trifluoroethoxy)carbonyl]amino}-1,3-thiazol-4-yl)acetic acid

  • MDL number MFCD13217733
  • Cas Number : 1306644-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182...184
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78200

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78200
Availability details for
Quantity
  • Availability
  • Add to Cart