ENA321765810 Enamine

3-(pyridin-3-yl)butanoic acid

  • MDL number MFCD11172879
  • Cas Number : 916262-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78208
Availability details for
Quantity
  • Availability
  • Add to Cart