ENA321765820 Enamine

5-bromo-2-(carbamoylamino)benzoic acid

  • MDL number MFCD16666288
  • Cas Number : 861791-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77932
Availability details for
Quantity
  • Availability
  • Add to Cart