ENA321765826 Enamine

2-{[(4-chlorophenyl)methyl]amino}acetic acid hydrochloride

  • MDL number MFCD17430341
  • Cas Number : 90562-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-217
Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78292
Availability details for
Quantity
  • Availability
  • Add to Cart