ENA321765830 Enamine

thiane-3-carboxylic acid

  • MDL number MFCD14584577
  • Cas Number : 161404-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76...78
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77779

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-77779
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart