ENA321765831 Enamine

4-chloro-7,8-difluoro-2-methylquinoline

  • MDL number MFCD12675034
  • Cas Number : 288151-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78305

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78305
Availability details for
Quantity
  • Availability
  • Add to Cart