ENA321765835 Enamine

3-methyl-4-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD16084562
  • Cas Number : 78726-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-50
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77937
Availability details for
Quantity
  • Availability
  • Add to Cart