ENA321765836 Enamine

2-(difluoromethoxy)-4-methoxybenzaldehyde

  • MDL number MFCD14607629
  • Cas Number : 1247613-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77938

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77938
Availability details for
Quantity
  • Availability
  • Add to Cart