ENA321765840 Enamine

4-cyclohexylbutan-2-amine

  • MDL number MFCD11655360
  • Cas Number : 791752-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78314
Availability details for
Quantity
  • Availability
  • Add to Cart