ENA321765842 Enamine

2-[(cyclohexylmethyl)sulfanyl]pyridine-3-carboxylic acid

  • MDL number MFCD11127471
  • Cas Number : 1019437-73-0
  • Molecular Weight : 251.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-78349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78349
Availability details for
Quantity
  • Availability
  • Add to Cart