ENA321765843 Enamine

2-[(4-hydroxyphenyl)formamido]acetamide

  • MDL number MFCD11130785
  • Cas Number : 1019364-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-78350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78350
Availability details for
Quantity
  • Availability
  • Add to Cart