ENA321765846 Enamine

[(1-amino-2-methylpropan-2-yl)oxy]benzene

  • MDL number MFCD11577413
  • Cas Number : 274686-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77941

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77941
Availability details for
Quantity
  • Availability
  • Add to Cart