ENA321765847 Enamine

N-methylquinolin-2-amine

  • MDL number MFCD10697835
  • Cas Number : 52430-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78354

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78354
Availability details for
Quantity
  • Availability
  • Add to Cart