ENA321765851 Enamine

1-(3-fluorophenyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD07374443
  • Cas Number : 248588-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78358
Availability details for
Quantity
  • Availability
  • Add to Cart