ENA321765865 Enamine

1-benzyl-2-methylpiperidin-4-one

  • MDL number MFCD00480711
  • Cas Number : 203661-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78382

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78382
Availability details for
Quantity
  • Availability
  • Add to Cart