ENA321765866 Enamine

ethyl 3-(ethylsulfanyl)butanoate

  • MDL number MFCD16088089
  • Cas Number : 90201-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78383

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78383
Availability details for
Quantity
  • Availability
  • Add to Cart