ENA321765875 Enamine

4-bromo-1-(2-methanesulfonylethyl)-1H-pyrazole

  • MDL number MFCD12780069
  • Cas Number : 1183252-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78417
Availability details for
Quantity
  • Availability
  • Add to Cart