ENA321765876 Enamine

2-[(4-bromophenyl)amino]acetic acid

  • MDL number MFCD00706053
  • Cas Number : 13370-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78418
Availability details for
Quantity
  • Availability
  • Add to Cart