ENA321765877 Enamine

(3-methyl-1-benzofuran-2-yl)methanol

  • MDL number MFCD15146108
  • Cas Number : 55581-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-77956

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77956
Availability details for
Quantity
  • Availability
  • Add to Cart