ENA321765881 Enamine

4-methyl-2-(1-methyl-1H-pyrazol-4-yl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD11207801
  • Cas Number : 1152599-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230...232
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78424

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78424
Availability details for
Quantity
  • Availability
  • Add to Cart