ENA321765882 Enamine

N-(1-cyano-1-methylethyl)benzamide

  • MDL number MFCD00835185
  • Cas Number : 54016-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78425
Availability details for
Quantity
  • Availability
  • Add to Cart