ENA321765885 Enamine

2-(cyclohexylmethyl)-3-methylbutanoic acid

  • MDL number MFCD12794610
  • Cas Number : 1247459-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78429
Availability details for
Quantity
  • Availability
  • Add to Cart