ENA321765888 Enamine

(2,2-dimethylpropyl)[2-(4-methoxyphenyl)ethyl]amine

  • MDL number MFCD14625026
  • Cas Number : 1226184-21-9
  • Molecular Weight : 221.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-78434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78434
Availability details for
Quantity
  • Availability
  • Add to Cart