ENA321765896 Enamine

ethyl 3-chloro-5H,6H,7H,8H-pyrido[4,3-c]pyridazine-6-carboxylate

  • MDL number MFCD07779187
  • Cas Number : 39715-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78445

Safety Information

msds https://www.enaminestore.com/catalog/EN300-78445
Availability details for
Quantity
  • Availability
  • Add to Cart