ENA321765897 Enamine

(cyclopropylmethyl)[(4-fluorophenyl)methyl]amine

  • MDL number MFCD11140258
  • Cas Number : 1019538-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78455
Availability details for
Quantity
  • Availability
  • Add to Cart