ENA321765902 Enamine

3-(4,5-dimethyl-2-oxo-2,3-dihydro-1,3-thiazol-3-yl)propanoic acid

  • MDL number MFCD09041361
  • Cas Number : 926214-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78460

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78460
Availability details for
Quantity
  • Availability
  • Add to Cart