ENA321765903 Enamine

2-(trifluoromethyl)-1,3-dioxaindan-2-amine

  • MDL number MFCD00725069
  • Cas Number : 146628-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77787

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77787
Availability details for
Quantity
  • Availability
  • Add to Cart