ENA321765907 Enamine

5-cyano-2-methylbenzene-1-sulfonamide

  • MDL number MFCD16679496
  • Cas Number : 1248118-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77961
Availability details for
Quantity
  • Availability
  • Add to Cart