ENA321765916 Enamine

4-[(2-hydroxyethyl)amino]-3-nitrobenzamide

  • MDL number MFCD12172421
  • Cas Number : 138377-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-78476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78476
Availability details for
Quantity
  • Availability
  • Add to Cart