ENA321765919 Enamine

3-(2-bromophenoxy)propan-1-ol

  • MDL number MFCD11614018
  • Cas Number : 52449-01-1
  • Molecular Weight : 231.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-77964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77964
Availability details for
Quantity
  • Availability
  • Add to Cart