ENA321765921 Enamine

3-(quinolin-4-yloxy)aniline

  • MDL number MFCD13438142
  • Cas Number : 1282557-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78512

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78512
Availability details for
Quantity
  • Availability
  • Add to Cart