ENA321765922 Enamine

(3-methyl-1-benzofuran-2-yl)methanamine

  • MDL number MFCD16556146
  • Cas Number : 3782-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77966

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77966
Availability details for
Quantity
  • Availability
  • Add to Cart