ENA321765929 Enamine

6-[(cyclohexylcarbamoyl)amino]hexanoic acid

  • MDL number MFCD09734446
  • Cas Number : 479413-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77810
Availability details for
Quantity
  • Availability
  • Add to Cart