ENA321765931 Enamine

1-(3-bromobenzenesulfonyl)-4-methylpiperazine

  • MDL number MFCD10000772
  • Cas Number : 486422-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78524
Availability details for
Quantity
  • Availability
  • Add to Cart