ENA321765932 Enamine

2-[(oxolan-2-yl)methoxy]ethan-1-amine

  • MDL number MFCD06248788
  • Cas Number : 34694-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78525
Availability details for
Quantity
  • Availability
  • Add to Cart