ENA321765935 Enamine

5-amino-1-methyl-3-(methylsulfanyl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD08236782
  • Cas Number : 1196151-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-78536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78536
Availability details for
Quantity
  • Availability
  • Add to Cart