ENA321765941 Enamine

1,2,3,4-tetrahydroisoquinoline-1-thione

  • MDL number MFCD00126731
  • Cas Number : 6552-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-102
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78542
Availability details for
Quantity
  • Availability
  • Add to Cart