ENA321765945 Enamine

bis(4-benzylpiperidine-1-carboximidamide); sulfuric acid

  • MDL number MFCD01701242
  • Cas Number : 59084-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2H2SO4
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77813
Availability details for
Quantity
  • Availability
  • Add to Cart