ENA321765946 Enamine

2-fluoro-4-methyl-5-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD12174289
  • Cas Number : 1155084-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78598
Availability details for
Quantity
  • Availability
  • Add to Cart