ENA321765947 Enamine

2-ethoxy-N'-hydroxypropanimidamide

  • Cas Number : 1251499-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-78599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78599
Availability details for
Quantity
  • Availability
  • Add to Cart