ENA321765948 Enamine

1-propoxyisoquinoline-3-carboxylic acid

  • MDL number MFCD11619125
  • Cas Number : 1094509-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77814

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77814
Availability details for
Quantity
  • Availability
  • Add to Cart